Learn More
Electron Microscopy Sciences Embedding Cassette with 6-Compartments, Tan
SDP
Catalog No. 5025068
Encompass_Preferred
Made from Acetal Polymer. These biopsy cassettes can hold up to six tissue specimens, each one placed in it's own compartment, numbered from 1 to 6 . Cover and base have over 3000 0.36 mm square openings. Cassettes keep specimen safely submerged in solvent and are totally resistant to the most histological solvents. The efficient flow-through slots maximize fluid exchange and insure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. Large labeling areas are located on three sides of the cassette. The anterior writing area is at a 45 degree angle, making the cassette, to some degree, able to be used with certain types of cassette labeling instruments.
Each case contains 42 dispenser boxes of 250 cassettes with covers assembled.
Color: Tan
Catalog No. 50-250-68 Supplier Electron Microscopy Sciences Supplier No. 70078T
Made from Acetal Polymer. These biopsy cassettes can hold up to six tissue specimens, each one placed in it's own compartment, numbered from 1 to 6 . Cover and base have over 3000 0.36 mm square openings. Cassettes keep specimen safely submerged in solvent and are totally resistant to the most histological solvents. The efficient flow-through slots maximize fluid exchange and insure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. Large labeling areas are located on three sides of the cassette. The anterior writing area is at a 45 degree angle, making the cassette, to some degree, able to be used with certain types of cassette labeling instruments.
Each case contains 42 dispenser boxes of 250 cassettes with covers assembled.
Color: Tan