Learn More
Electron Microscopy Sciences Process/Embedding Cassettes w/o Lids, Aqua
SDP
Catalog No. 5030287
Encompass_Preferred
Disposable plastic tissue cassettes are used for holding and identifying tissue samples in processing, embedding, and sectioning procedures. The cassettes fit securely in microtome chuck adapters. They are totally resistant to histological solvents. These cassettes are designed to accept standard metal lids and will also fit either the standard stainless steel base molds or the disposable plastic ones. Made from Acetal copolymer.
Color: Aqua
Catalog No. 50-302-87 Supplier Electron Microscopy Sciences Supplier No. 70025A
Disposable plastic tissue cassettes are used for holding and identifying tissue samples in processing, embedding, and sectioning procedures. The cassettes fit securely in microtome chuck adapters. They are totally resistant to histological solvents. These cassettes are designed to accept standard metal lids and will also fit either the standard stainless steel base molds or the disposable plastic ones. Made from Acetal copolymer.
Color: Aqua