Learn More
StatLab Embedding Cassettes
SDP
Catalog No. 22031877
Change view
Click to view available options
Quantity:
1500/Cs.
Catalog No. Quantity
22-031-877 1500/Cs.
1 options
Catalog No. 22-031-877 Supplier StatLab Supplier No. M4922
Add to Cart
Add to Cart
Embedding Cassettes - Routine Embedding Cassettes
  • Unique front-hinged design with a rear tab that allows for unobstructed access to the front labeling surface
  • Hinged lid, rear tab
  • Bulk packed 1500/Cs.
  • 45° labeling surface that is compatible with inkjet and thermal transfer printers
Specifications
Quantity 1500/Cs.
Color White
Product Type Embedding Cassette