Learn More
StatLab Embedding Cassettes
SDP
Catalog No. 21033416
Change view
Click to view available options
Color:
Green
Lilac
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-033-416 Lilac
21-033-417 White
21-033-418 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-033-416 Supplier StatLab Supplier No. M48010
Only null left
Add to Cart
Add to Cart
Designed to be used with re-usable metal lid SKU PCLID
  • No lids
  • 45-degree labeling surface
  • Taped, CS750, Bulk packed, CS1500
Specifications
Color Lilac
Product Type Embedding Cassette