Learn More
Leica Biosystems Leica Biosystems EM-400 Embedding Medium Paraffin - 2 lb bags, 10 per case
Catalog No. NC3037525
Encompass_Preferred
EM-400 embedding medium paraffin is a purified paraffin blend with synthetic polymers formulated for histology tissue embedding and routine microtomy. It minimizes compression during sectioning, supports thin sectioning, and remains stable at elevated temperatures to help reduce artifactual tissue distortion.
  • Medium melting point (56-57 °C) for reliable embedding.
  • Recommended water bath temperature 37-42 °C to help preserve tissue morphology.
  • Packaged as 10 x 2 lb bags for laboratory convenience.
  • Formulated to minimize compression and support 1-2 μm sectioning.
  • Stable at elevated temperatures to reduce block variability.
Catalog No. NC3037525 Supplier Leica Biosystems Supplier No. 3801320
EM-400 embedding medium paraffin is a purified paraffin blend with synthetic polymers formulated for histology tissue embedding and routine microtomy. It minimizes compression during sectioning, supports thin sectioning, and remains stable at elevated temperatures to help reduce artifactual tissue distortion.
  • Medium melting point (56-57 °C) for reliable embedding.
  • Recommended water bath temperature 37-42 °C to help preserve tissue morphology.
  • Packaged as 10 x 2 lb bags for laboratory convenience.
  • Formulated to minimize compression and support 1-2 μm sectioning.
  • Stable at elevated temperatures to reduce block variability.