Learn More
Electron Microscopy Sciences Embedding Mold, Dykstra
SDP
Catalog No. 50930541
Encompass_Preferred
EMS Embedding Mold is a new mold designed with Michael J. Dykstra Ph.D. from North Carolina State University Department of Veterinary Medicine. This Mold produces blocks of the best dimension to be held securely by Reichert/Leica Ultracut stepped microtome chucks. There are 24 cavities and the depth of each cavity is 4 mm which will prevent breakage of the blocks during razor blade trimming. Mold Measurements: 3 3/8" x 3" (85 x 77 mm. Cavity Measurements: 15 mmx 7 mm x 4 mm deep
Catalog No. 50-930-541 Supplier Electron Microscopy Sciences Supplier No. 70907
EMS Embedding Mold is a new mold designed with Michael J. Dykstra Ph.D. from North Carolina State University Department of Veterinary Medicine. This Mold produces blocks of the best dimension to be held securely by Reichert/Leica Ultracut stepped microtome chucks. There are 24 cavities and the depth of each cavity is 4 mm which will prevent breakage of the blocks during razor blade trimming. Mold Measurements: 3 3/8" x 3" (85 x 77 mm. Cavity Measurements: 15 mmx 7 mm x 4 mm deep