Learn More
Electron Microscopy Sciences Peel Away Disposable Embedding Mold Square Shape, 22 x 22 mm
SDP
Catalog No. NC1513275
Encompass_Preferred
Peel Away Disposable Embedding Molds are used for plastic or paraffin embedding. No block trimming needed. Inside fingers hold ID slip which becomes embedded in the block for positive permanent identification.
Size: 22 x 22 mm
Catalog No. NC1513275 Supplier Electron Microscopy Sciences Supplier No. 70182
Peel Away Disposable Embedding Molds are used for plastic or paraffin embedding. No block trimming needed. Inside fingers hold ID slip which becomes embedded in the block for positive permanent identification.
Size: 22 x 22 mm