Learn More
Ted Pella Inc EMBEDDING MOLDS 22X22 288/CS
SDP
Catalog No. NC9719245
Encompass
Disposable histology Mold, Peel-A-Way;, 22mm Square, 20mm Deep, Useful for Most Plastic Embedding Media and for Paraffin Waxes, Especially Suited for Paraffins, May be Used Under Anaerobic Conditions
Catalog No. NC9719245 Supplier Ted Pella Inc Supplier No. 27110
Disposable histology Mold, Peel-A-Way;, 22mm Square, 20mm Deep, Useful for Most Plastic Embedding Media and for Paraffin Waxes, Especially Suited for Paraffins, May be Used Under Anaerobic Conditions