Learn More
Electron Microscopy Sciences Embedding Ring, Yellow
SDP
Catalog No. 5025736
Encompass_Preferred
Molded from the highest quality plastic, our Embedding Rings fit most base molds. The large writing surface further eases identification and handling. The base measures 1 5/8" x 1 1/8" and the ring measures 1" x 1" (25 x 25 mm).
Catalog No. 50-257-36 Supplier Electron Microscopy Sciences Supplier No. 62350Y
Molded from the highest quality plastic, our Embedding Rings fit most base molds. The large writing surface further eases identification and handling. The base measures 1 5/8" x 1 1/8" and the ring measures 1" x 1" (25 x 25 mm).