Learn More
Cayman Chemical Embelin, Purity: ≥95%, CAS: 550-24-3, 50 mg
SDP
Catalog No. 502952678
Encompass_Preferred
A benzoquinone with diverse biological activities; inhibits viral replication of the influenza A subtype H1N1 and the influenza B subtype Victoria-like in MDCK cells (IC50s = 0.3 and 0.2 µM, respectively); an inhibitor of XIAP (IC50 = 4.1 µM in a fluorescence polarization assay; induces apoptosis and increases caspase-9 levels in PC3 cells at 20-50 µM; completely inhibits TNF-α-induced NF-κB and IKK activation and inhibits TNF-α-induced IκBα phosphorylation in KBM5 cells at 50 µM; inhibits TNF-α-induced IκBα phosphorylation at 50 µM; active against the Gram-positive bacteria B. subtilis, S. aureus, and M. luteus (MICs = 20, 20, and 25 µg/ml, respectively) and the Gram-negative bacteria P. aeruginosa, K. pneumoniae, and E. coli (MICs = 25, 25, and 45 µg/ml, respectively)
Catalog No. 50-295-2678 Supplier Cayman Chemical Supplier No. 1183850 MG
A benzoquinone with diverse biological activities; inhibits viral replication of the influenza A subtype H1N1 and the influenza B subtype Victoria-like in MDCK cells (IC50s = 0.3 and 0.2 µM, respectively); an inhibitor of XIAP (IC50 = 4.1 µM in a fluorescence polarization assay; induces apoptosis and increases caspase-9 levels in PC3 cells at 20-50 µM; completely inhibits TNF-α-induced NF-κB and IKK activation and inhibits TNF-α-induced IκBα phosphorylation in KBM5 cells at 50 µM; inhibits TNF-α-induced IκBα phosphorylation at 50 µM; active against the Gram-positive bacteria B. subtilis, S. aureus, and M. luteus (MICs = 20, 20, and 25 µg/ml, respectively) and the Gram-negative bacteria P. aeruginosa, K. pneumoniae, and E. coli (MICs = 25, 25, and 45 µg/ml, respectively)