Learn More
Active Motif Embelin
SDP
Catalog No. NC1841438
Encompass
Active Motif's Embelin inhibits the lysine acetyltransferase PCAF in vitro and in vivo (in vitro IC50 = 7.2 uM). It inhibits H3K9 acetylation in mouse liver tissue while leaving levels of H3K14, H4K8, and H4K12 unchanged. It also inhibits X-linked inhibitor of apoptosis (XIAP) (IC50 = 4.1 uM, competing with Smac peptide). Embelin to the BIR3 domain, preventing XIAP interaction with caspase-9 and Smac. It induces apoptosis and alters gene expression profiles in breast cancer cells. Embelin possesses anti-inflammatory and analgesic activity in vivo, and it is cell permeable.
Catalog No. NC1841438 Supplier Active Motif Supplier No. 14118
Active Motif's Embelin inhibits the lysine acetyltransferase PCAF in vitro and in vivo (in vitro IC50 = 7.2 uM). It inhibits H3K9 acetylation in mouse liver tissue while leaving levels of H3K14, H4K8, and H4K12 unchanged. It also inhibits X-linked inhibitor of apoptosis (XIAP) (IC50 = 4.1 uM, competing with Smac peptide). Embelin to the BIR3 domain, preventing XIAP interaction with caspase-9 and Smac. It induces apoptosis and alters gene expression profiles in breast cancer cells. Embelin possesses anti-inflammatory and analgesic activity in vivo, and it is cell permeable.