Learn More
Biotium EMBER500 RNA PRESTAIN 250UL
Catalog No. NC2103165
Encompass_Preferred
EMBER500 RNA Prestain Loading Dye is a convenient 2X RNA loading dye for single-step denaturing, staining, and loading of RNA for gel analysis. EMBER500™ dye stains RNA and DNA with much brighter signal than ethidium bromide, and can be used with both UV transilluminators and blue LED gel imagers.
Catalog No. NC2103165 Supplier Biotium Supplier No. 41032250UL
EMBER500 RNA Prestain Loading Dye is a convenient 2X RNA loading dye for single-step denaturing, staining, and loading of RNA for gel analysis. EMBER500™ dye stains RNA and DNA with much brighter signal than ethidium bromide, and can be used with both UV transilluminators and blue LED gel imagers.