Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Embramine hydrochloride | 13977-28-1 | 384.74 | 10 MG
SDP

Catalog No. 5000448402
Encompass_Preferred

Embramine hydrochloride is a monoethanolamine used as an antihistamine and anticholinergic, with antihistamine and anticholinergic properties.

  • Used as an antihistamine and anticholinergic
  • Molecular weight: 384.74
  • Formula: C18H23BrClNO
  • Data sheet and handling instructions available
  • For research use only; not for sale to patients

Catalog No. 50-004-48402 Supplier Medchemexpress LLC Supplier No. HYU00132A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Embramine hydrochloride is a monoethanolamine used as an antihistamine and anticholinergic, with antihistamine and anticholinergic properties.

  • Used as an antihistamine and anticholinergic
  • Molecular weight: 384.74
  • Formula: C18H23BrClNO
  • Data sheet and handling instructions available
  • For research use only; not for sale to patients

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.