Learn More
NEXPRING HEALTH EMBRYO CULTURE OIL 100ML
Catalog No. NC9938801
Encompass
Oil for Embryo Culture. 100ml. Oil for Embryo Culture is a sterile light mineral oil intended for use as an overlay when culturing in reduced volumes of media to prevent evaporation, and to protect the media from changes in osmolality and pH.
Catalog No. NC9938801 Supplier NEXPRING HEALTH Supplier No. 9305100ML
Oil for Embryo Culture. 100ml. Oil for Embryo Culture is a sterile light mineral oil intended for use as an overlay when culturing in reduced volumes of media to prevent evaporation, and to protect the media from changes in osmolality and pH.