Learn More
Electron Microscopy Sciences Embryo Dish, 45 mm O.D
SDP
Catalog No. 50930384
Encompass_Preferred
These dishes are molded from clear glass and come with a glass lid for the viewing or staining of free-floating specimens.
Catalog No. 50-930-384 Supplier Electron Microscopy Sciences Supplier No. 7054345
These dishes are molded from clear glass and come with a glass lid for the viewing or staining of free-floating specimens.