Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ EmbryoMax™ PMEF, Hygro Resistant, Strain C57/BL6, Passage 3

Catalog No. PMEFH Shop All MilliporeSigma Products
Change view
Click to view available options
Cell Type:
Primary MEF
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Type
PMEFH Primary MEF
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PMEFH Supplier MilliporeSigma™ Supplier No. PMEFH
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For stem cell culture applications

  • Fibroblasts can be directly plated onto gelatin coated culture vessels
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -80°C
Cell Type Primary MEF
For Use With (Application) Stem cell culture
Quantity 5 vials

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.