Learn More
ABclonal Technology EMC8 Rabbit pAb
SDP
Catalog No. 5000104704
Encompass_Preferred
Contributes to membrane insertase activity. Involved in protein insertion into ER membrane by stop-transfer membrane-anchor sequence and tail-anchored membrane protein insertion into ER membrane. Located in cytosol. Part of EMC complex.
Catalog No. 50-001-04704 Supplier ABclonal Technology Supplier No. A148381000UL
Contributes to membrane insertase activity. Involved in protein insertion into ER membrane by stop-transfer membrane-anchor sequence and tail-anchored membrane protein insertion into ER membrane. Located in cytosol. Part of EMC complex.