Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ β1,2-Xylosidase, Xanthomonas sp., Protein

Catalog No. 684274500MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mIU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-427-4500MI 500 mIU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-427-4500MI Supplier MilliporeSigma™ Supplier No. 684274500MIU
Add to Cart
Add to Cart
Native β1,2-Xylosidase from Xanthomonas species. Catalyzes the hydrolysis of terminal xylose β1,2-linked to the β-mannose of N-linked oligosaccharides. Enzyme requires removal of α1,3-linked mannose. Enzyme requires 5mM calcium chloride in reaction buffer for full activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 25mM NaCl, 20mM Tris-HCl, pH 7.5.
Quantity 500 mIU
Source Xanthomonas sp.
Immunogen β1,2-Xylosidase
Storage Requirements Following initial use, refrigerate (4°C).
Regulatory Status RUO
Recombinant Native
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.