Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ α1-3,6-Galactosidase, Recombinant Protein, E. coli

Catalog No. 34579725U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-579-725U 25 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-579-725U Supplier MilliporeSigma™ Supplier No. 34579725U
Add to Cart
Add to Cart
Recombinant α1-3,6-galactosidase expressed in E. coli. Catalyzes the hydrolysis of α1,3-linked and α1,6-linked, non-reducing terminal galactose from complex carbohydrates and glycoproteins. Particularly efficient in removing α-linked galactose under conditions where the pH must be neutral or above, as with live cells.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 50mM sodium phosphate buffer, pH 7.5.
Quantity 25 U
Source E. coli
Immunogen α1-3,6-Galactosidase
Regulatory Status RUO
Recombinant Recombinant
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.