Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ HRV 3C Protease

Catalog No. 714933 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-493-3 500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-493-3 Supplier MilliporeSigma™ Supplier No. 714933
Add to Cart
Add to Cart

Highly efficient fusion tag removal at low temperature

  • Recombinant type 14 3C protease from human rhinovirus (HRV 3C) is a recombinant restriction-grade protease
  • HRV 3C is a highly purified recombinant 6XHis-fusion protein, that recognizes the same cleavage site as the native enzyme: LeuGluValLeuPheGln↓GlyPro
  • The small, 22-kDa size of the protease, optimal activity at 4°C, high specificity, and His•Tag™ fusion make HRV 3C protease an ideal choice for rapid removal of fusion tags
  • The pET expression vectors pET-47b(+) through pET-50b(+) incorporate the HRV 3C protease cleavage site in combinations with His•Tag, S•Tag™, thioredoxin (Trx•Tag™), glutathione-S-transferase (GST•Tag™), and NusA (Nus•Tag™) sequences
  • The combination of pET-47b(+) to pET-50b(+) vectors for expression, HRV 3C protease for fusion tag cleavage, and Ni-NTA His•Bind™ metal affinity chromatography for protein purification as well as protease and fusion tag removal allows production of recombinant proteins free of vector-encoded sequences
  • Other expression vectors, including the pET-52b(+) vector, the InsectDirect™ pIEx™-9 vector, and the multisystem pTriEx™-6 vector, all incorporate the HRV 3C protease cleavage site in combination with the Strep•Tag™ II and His•Tag sequences
  • Supplied as: 1.8-3.0U/μl in 50mM Tris-HCl (pH 8.0), 150mM NaCl, 1mM EDTA, 0.5mM THP, and 50% glycerol
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 50mM Tris-HCl (pH 8.0), 150mM NaCl, 1mM EDTA, 0.5mM THP, and 50% glycerol
Content And Storage -20°C
Quantity 500 U
Product Type Protease
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.