Learn More
MilliporeSigma™ 96-well V-bottom Collection Plate
Catalog No. MSCPNPP00 Shop All MilliporeSigma Products
Change view
Click to view available options
No. per Pack:
100
Catalog No. No. per Pack
MSCPNPP00 100
1 options
Catalog No. MSCPNPP00 Supplier MilliporeSigma™ Supplier No. MSCPNPP00
Add to Cart
Add to Cart
96-well V-bottom collection plate
TRUSTED_SUSTAINABILITY
Specifications
Array 8 x 12
Color Clear
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
No. per Pack 100
Quantity 100/Pk.