Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ αA-Interferon, Human, Recombinant Protein, E. coli

Catalog No. 4072945MU Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5mU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-729-45MU 5mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-729-45MU Supplier MilliporeSigma™ Supplier No. 4072945MU
Only null left
Add to Cart
Add to Cart
Recombinant, human αA-interferon expressed in E. coli. Antiviral, anti-neoplastic agent. αA-Interferon exhibits therapeutic potential with chronic viral hepatitis, polycythemia vera, and various malignancies. It enhances natural killer cell activity, suppresses bone marrow function, and induces apoptotic cell death in non-melanoma skin cancer. NOTE: 1MU = 1,000,000 units.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In PBS containing 0.1% BSA.
Preparation Method Dilute only in buffers containing protein such as 0.1% BSA.
Purification Method SDS-PAGE
Quantity 5mU
Source E. coli
Immunogen αA-Interferon
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.