Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Acetyl-Histone H3 (Lys14)α, Rabbit Monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710051MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-100-51MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-51MI Supplier MilliporeSigma™ Supplier No. 1710051

Rabbit Monoclonal Antibody

Specifically detects Acetyl-Histone H3 (Lys14)α Clone: in Human, Rat samples, and it is validated for ChIP, ChIP-seq, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blotting

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the 'beads on a string' structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetyl-Histone H3 (Lys14)α
Applications ChIP sequencing (ChIP-seq), Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-Acetyl-Histone H3 (Lys14) (rabbit monoclonal). One vial containing 50μL of unpurified Monoclonal in buffer containing 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, with 40% Glycerol, 0.01% sodium azide and 0.05% BSA. Nornal Rabbit IgG. One vial containing 75μL of normal rabbit IgG. ChIP Primers, c-myc promoter. One vial containing 75μL of 5μM of each primer specific for human c-myc promoter. FOR: CCC AAA AAA AGG CAC GGA A; REV: TAT TGG AAA TGC GGT CAT GC
Gene Accession No. P84243
Gene Symbols H3.3A, H3.3B, H3F3, MGC87782, MGC87783, OTTHUMP00000035618, OTTHUMP00000035619, OTTHUMP00000035621
Host Species Rabbit
Immunogen Acetyl-specific synthetic peptide corres-ponding to residues around Lys14 of Histone H3.
Purification Method Unpurified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003484.1
Test Specificity Recognizes Histone H3 acetylated on Lys14.
Includes This ChIPAb+ Acetyl-Histone H3 (Lys14) -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Rat
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.