Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Acetyl-Histone H3 (Lys27)α, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17683MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-683-MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-683-MI Supplier MilliporeSigma™ Supplier No. 17683
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects Acetyl-Histone H3 (Lys27)α Clone: in Vertebrate samples, and it is validated for Chromatin Immunoprecipitation, Western Blotting

Lysine acetylation is a dynamic, reversible and tightly regulated protein and histone modification that plays a major role in chromatin remodeling and in the regulation of gene expression in various cellular functions. Histone acetylation is often associated with transcriptional activation and acetylation of H3K27 is found at active enhancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetyl-Histone H3 (Lys27)α
Applications ChIP Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-acetyl-Histone H3 (Lys27) (mouse monoclonal IgG1, Clone CMA309). One vial containing 50μg of protein G purified antibody in 50μL PBS containing 0.05% sodium. Normal Mouse IgG. Two vials containing 25μg purified Mouse IgG in 25μL storage buffer containing 0.1% sodium azide. ChIP Primers, RPL10 Promoter. One vial containing 75μL of 5μM of each primer specific for the promoter region of human RPL10. FOR: ACC CGT CTT CGA CAG GAC T; REV: GGA ACG GAA GAC GAG AAC AG
Gene Accession No. Q66I33
Host Species Mouse
Immunogen The acetyl-histone H3 (Lys27) purified antibody is made against a synthetic peptide (acetylated at Lys27) corresponding to amino acids 19-37 of histone H3.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NM_003493
Test Specificity Recognizes histone H3, Mr 17kDa, acetylated at Lys27.
Includes Acetyl-Histone H3 (Lys27) ChIP validated Antibody and Primer set including the ChIP-grade antibody and the specific control PCR primers used for chromatin immunoprecipitation of H3K27Ac.
Target Species Vertebrates
Content And Storage −20°C for up to 12 months; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.