Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Acetyl-Histone H3 (Lys9/18), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710241MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-102-41MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-41MI Supplier MilliporeSigma™ Supplier No. 1710241
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifically detects Acetyl-Histone H3 (Lys9/18) Clone: in Bovine, Human, Mouse, S. cerevisiae, Yeast samples, and it is validated for Chromatin Immunoprecipitation, Western Blotting

Histone H3 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H3 is involved with the structure of the nucleosomes of the 'beads on a string' structure. Acetylation of histone H3 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetyl-Histone H3 (Lys9/18)
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-Acetyl-Histone H3 (Lys9/18) (rabbit polyclonal). One vial containing 25μL of rabbit antiserum containing 0.05% sodium azide before the addition of glycerol to 30%. Normal Rabbit Serum. One vial containing 25μL of antiserum containing 0.05% sodium azide. Control Primers, human GAPDH promoter. One vial containing 75μL of 5μM of each primer specific for human GAPDH. FOR: TAC TAG CGG TTT TAC GGG CG; REV: TCG AAC AGG AGG AGC AGA GAG CGA
Gene Accession No. Q16695
Gene Symbols H3F3A, MGC87783, H3.3A, MGC87782, H3F3, H3.3B, H3F3B
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide (..ARAcKSTGGKAPRAcKQL..) in which AcK corresponds to acetyl-lysine at residue 9 and 18 of human Histone H3.
Purification Method Unpurified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NM_003493
Test Specificity Recognizes Histone H3 acetylated on lysines 9 and 18.
Includes This ChIPAb+ Acetyl-Histone H3 (Lys9/18) -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Bovine, Human, Mouse, S. cerevisiae, Yeast
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.