Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Acetyl-Histone H3 (Lys9) Serum, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17609MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-609-MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-609-MI Supplier MilliporeSigma™ Supplier No. 17609
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifically detects Acetyl-Histone H3 (Lys9) Serum Clone: in Human, Mammalia, Mouse samples, and it is validated for ChIP, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetyl-Histone H3 (Lys9) Serum
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-Acetyl-Histone H3 (Lys9) (rabbit polyclonal serum). One vial containing 25μL of antiserum containing 0.05% sodium azide before the addition of glycerol to 30%.
Normal Rabbit Serum. One vial containing 25μL antiserum containing 0.05% sodium azide.
Control Primers. One vial containing 75μL of 5μM of each primer specific for for human GAPDH. FOR: TAC TAG CGG TTT TAC GGG CG REV: TCG AAC AGG AGG AGC AGA GAG CGA
Gene Accession No. Q66I33
Gene Symbols H3F3A, H3F3B, H3F3
Host Species Rabbit
Immunogen The acetyl-histone H3 (Lys9) antiserum is made against a peptide corresponding to amino acids 4-14 of yeast histone H3 acetylated on lysine 9.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NM_003493
Test Specificity Acetyl-Histone H3 (Lys9)
Includes This ChIPAb+ Acetyl-Histone H3 (Lys9) Serum -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mammalia, Mouse
Content And Storage −20°C from date of receipt
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.