Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Acetyl-Histone H3, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17615MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
125 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-615-MI 125 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-615-MI Supplier MilliporeSigma™ Supplier No. 17615
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifically detects Acetyl-Histone H3 Clone: in Human, Mouse, Tetrahymena samples, and it is validated for ChIP, Western Blotting

Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. Acetylation is generally linked to gene activation. Acetylation on Lys-10 (H3K9ac) impairs methylation at Arg-9 (H3R8me2s). Acetylation on Lys-19 (H3K18ac) and Lys-24 (H3K24ac) favors methylation at Arg-18 (H3R17me).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetyl-Histone H3
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-Acetyl-Histone H3 (rabbit polyclonal IgG). One vial containing 125μg of protein A purified antibody in 125μL of storage buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide. Normal Rabbit IgG. One vial containing 125μg of normal rabbit IgG in 125μL volume. Control Primers. One vial containing 75μL of 5μM of each primer specific for for human GAPDH. FOR: TAC TAG CGG TTT TAC GGG CG; REV: TCG AAC AGG AGG AGC AGA GAG CGA
Gene Accession No. P68431
Host Species Rabbit
Immunogen The acetyl-histone H3 antibody is made against a peptide corresponding to amino acids 1-20 of Tetrahymena histone H3 ARTKQTAR[K]STGG[K]APRKQL(C) where [K] is acetylated)
Purification Method Protein A purified
Quantity 125 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003484
Test Specificity Recognizes acetyl-histone H3
Includes Acetyl-Histone H3 ChIP validated Antibody and Primer set including the ChIP-grade antibody and the specific control PCR primers used for chromatin immunoprecipitation of H3Ac.
Target Species Human, Mouse, Tetrahymena species
Content And Storage −20°C from date of receipt
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.