Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Acetyl-Histone H4 (Lys12)α, Rabbit Monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710121MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-101-21MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-101-21MI Supplier MilliporeSigma™ Supplier No. 1710121
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifically detects Acetyl-Histone H4 (Lys12)α Clone: in Human, Vertebrate samples, and it is validated for ChIP, Functional Assay, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetyl-Histone H4 (Lys12)α
Applications ChIP Assay, Functional Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-Acetyl-Histone H4 (Lys12) (rabbit monoclonal), . One vial containing 50μL of rabbit monoclonal IgG cell culture supernatant in 0.1% sodium azide. Negative Control Supernatant, . One vial containing 100μL of cultured supernatant in 0.05% sodium azide. ChIP Primers, human GAPDH coding region. One vial containing 75μL of 5μM each primer specific for human GAPDH coding region. FOR: GGC TCC CAC CTT TCT CAT CC; REV: GGC CAT CCA CAG TCT TCT GG
Gene Accession No. P62805
Gene Symbols HIST1H4A, H4/J, HIST1H4F, HIST1H4L, H4FN, H4FA, H4FH, HIST1H4H, H4F2, H4FM, H4/A, H4FK, H4FG, HIST2H4, H4FB, H4/K, HIST1H4I, H4/N, HIST1H4B, H4FE, H4/M, H4/a, HIST1H4E, HIST4H4, HIST2H4A, H4FC, H4FI, H4/H, HIST1H4C, H4/G, HIST1H4K, H4FJ, H4FD, H4/I, H4/B, H4/D, H4/C, HIST1H4J, HIST1H4D, H4/E
Host Species Rabbit
Immunogen Peptide corresponding to Histone H4 containing the sequence [GLG-AcK-GGA] on which Lys12 is acetylated.
Purification Method Protein A purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NM_003538.3
Test Specificity This antibody detects histone H4 acetylated at Lys12.
Includes This ChIPAb+ Acetyl-Histone H4 (Lys12) -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Vertebrates
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.