Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Acetyl-Histone H4 (Lys16), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710101MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-101-01MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-101-01MI Supplier MilliporeSigma™ Supplier No. 1710101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifically detects Acetyl-Histone H4 (Lys16) Clone: in Human, Vertebrate samples, and it is validated for ChIP, Immunoprecipitation, Western Blotting

Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H4 is involved with the structure of the nucleosomes of the 'beads on a string' structure. Acetylation of histone H4 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetyl-Histone H4 (Lys16)
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-Acetyl-Histone H4 (Lys16) (rabbit polyclonal). One vial containing 50μL of affinity purified rabbit polyclonal serum in buffer containing 0.1 M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% sodium azide. Normal Rabbit IgG. One vial containing 125μg of rabbit IgG in 125μL of storage buffer containing 0.05% sodium azide. ChIP Primers, RPL10 promoter. One vial containing 75μL of 5μM of each primer specific for the promoter region of human RPL10. FOR: ACC CGT CTT CGA CAG GAC T; REV: GGA ACG GAA GAC GAG AAC AG
Gene Accession No. P62805
Gene Symbols HIST1H4A, H4/J, HIST1H4F, HIST1H4L, H4FN, H4FA, H4FH, HIST1H4H, H4F2, H4FM, H4/A, H4FK, H4FG, HIST2H4, H4FB, H4/K, HIST1H4I, H4/N, HIST1H4B, H4FE, H4/M, H4/a, HIST1H4E, HIST4H4, HIST2H4A, H4FC, H4FI, H4/H, HIST1H4C, H4/G, HIST1H4K, H4FJ, H4FD, H4/I, H4/B, H4/D, H4/C, HIST1H4J, HIST1H4D, H4/E
Host Species Rabbit
Immunogen KLH-conjugated Histone H4 corresponding to the N-terminus region at and around acetylated Lys16.
Purification Method Affinity Purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_778224.1
Test Specificity This antibody detects histone H4 acetylated at Lys16.
Includes This ChIPAb+ Acetyl-Histone H4 (Lys16) -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Vertebrates
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.