Learn More
MilliporeSigma™ Activation Code for Millitrack™ Software
Catalog No. ACTIC0M04 Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
RiOs™ and Elix™ Millitrack Software
Catalog No. For Use With (Equipment)
ACTIC0M04 RiOs™ and Elix™ Millitrack Software
1 options
Catalog No. ACTIC0M04 Supplier MilliporeSigma™ Supplier No. ACTIC0M04
Add to Cart
Add to Cart
Provides data management, archiving and remote access capabilities
  • For RiOs™ and/or Elix™ software
  • User-friendly installation
  • Millitrack Software helps in network settings adjustment and familiarization
  • One user license access per lab water system
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) RiOs™ and Elix™ Millitrack Software