Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Aerosol Adapter, Luer Slip to 1/4 in.– 3/8 in. ID Hose

Catalog No. XX6200004 Shop All MilliporeSigma Products
Change view
Click to view available options
Material:
Stainless Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
XX6200004 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. XX6200004 Supplier MilliporeSigma™ Supplier No. XX6200004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adapts vacuum source to filter holders

Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
TRUSTED_SUSTAINABILITY

Specifications

Connectors 0.25 to 0.37 in. I.D. Hose
Material Stainless Steel
For Use With (Application) To connect the vacuum source to any millipore aerosol analysis monitor or Swinnex™ 47 mm membrane filter holder
Includes Aerosol Adapter, Luer slip to 1/4 in.- 3/8 in. ID hose, stainless steel
Type Environmental Test Kits or Supplies
Diameter (English) Inside 0.25 in.
Diameter (Metric) Inside 0.63 cm
For Use With (Equipment) Millipore Aerosol Analysis Monitor or Swinnex™ 47 mm Membrane Filter Holder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.