Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Amicon™ Bioseparations Stirred Cells†

Catalog No. 5123 Shop All MilliporeSigma Products
Change view
Click to view available options
Model 8200; Filter dia.: 63.5mm; Filtrate holdup vol.: 1200μL; Housing: 9L x 9W x 12.8cmH
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
5123 Model 8200; Filter dia.: 63.5mm; Filtrate holdup vol.: 1200μL; Housing: 9L x 9W x 12.8cmH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5123 Supplier MilliporeSigma™ Supplier No. 5123

missing translation for 'validMainframeMsg'

Includes

Connection for 0.25 in. dia. (6.4mm) pressure tubing; max. operating pressure, 75psi (5.3 bar).

Requires

Magnetic stirrer required for operation.

Concentrate, desalt, and purify aqueous biological solutions

Process from 3mL up to 400mL of biological solutions using either concentration or diafiltration operating modes.

  • Sturdy construction; autoclavable Consist of clear pressurized vessel with integral magnetic stirring bar, nylon cap and tube fitting assembly, polysulfone body and membrane holder, acetal/polysulfone stirrer, silicone O-rings, and black aluminum stand
Concentration Mode
  • Gas pressure is applied directly to ultrafiltration cell Solutes above membrane molecular weight cutoff are retained in cell, while water and solutes below cutoff pass into filtrate
Diafiltration Mode
    Ultrafiltration cell is connected to auxiliary reservoir containing diafiltrate solution; reservoir is then gas pressurized Fluid volume and macrosolute concentration remain constant as filtrate is replaced by diafiltrate solution Provides a simple means for rapid microsolute exchange
  • Typically used as a substitute for dialysis

Applications:

Perform concentration, desalting, recovery or exclusion; Recover monoclonal antibodies and growth factors from tissue culture supernatants; Concentration and buffer exchange of chromatography fractions

Order Info

Use with Millipore Ultrafiltration Membranes.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Concentration 0.05 to 10g%
Material Nylon cap and tube fitting assembly, Polysulfone cylinder and membrane holder, Acetal and Polysulfone stirring assembly, Silicone Rubber O-rings, Polyethelene pressure tube, Tygon tubing filtrate tube, Anodized Aluminum or Nylon retaining stand
Filtration Area 28.7 cm2
For Use With (Application) Protein purification, protein concentration, protein desalting
Includes Clear pressurized vessel with integral magnetic stirring bar, nylon cap and tube fitting assembly, polysulfone body and membrane holder, acetal/polysulfone stirrer, silicone O-rings and black aluminum stand, connections for 0.25 in. dia. pressure tubing
Model 8200
Volume (English) 0.16 to 6.7 oz.
Volume (Metric) 5 to 200 mL
Volume (Metric) Hold Up 1200 μL
Dimensions (L x W x H) 3.5 x 3.5 x 5.03 in. (9 x 9 x 12.8cm)
Length (English) 3.5 in.
Length (Metric) 9 cm
Width (English) 3.5 in.
Width (Metric) 9 cm
Height (English) 5.03 in.
Height (Metric) 12.8 cm
Diameter (English) 2.5 in.
Diameter (Metric) 6.35 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.