Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Amicon™ Pro Affinity Concentration Kit Protein A with Amicon™ Ultra-0.5 Device

Catalog No. ACK5030PA Shop All MilliporeSigma Products
Change view
Click to view available options
MWCO:
100 kDa
30 kDa
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO
ACK5030PA 30 kDa
ACK5100PA 100 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ACK5030PA Supplier MilliporeSigma™ Supplier No. ACK5030PA

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Includes

  • 12/pk Amicon Pro
  • 12/pk Amicon Ultra
  • 3mL Protein A resin (Binding capacity: 20mg human IgG/mL agarose)
  • 2 X 66mL Bind/Wash Buffer
  • 12mL Elution buffer
  • 2.5mL Neutralization buffer

Amicon Pro Affinity Concentration Kit Protein A - A Centrifugal Tool for purifying and concentrating Antibodies.

This kit includes 12/pk Amicon Pro, 12/pk Amicon Ultra NMWCO, 3mL Protein A resin (Binding capacity: 20mg human IgG/ml agarose), 2 X 66ml Bind/Wash Buffer, 12mL Elution buffer, 2.5mL Neutralization buffer

Order Info

Reagents available. (See Cat. No. ACR5000PA)

TRUSTED_SUSTAINABILITY

Specifications

Color Clear with Pink Cap
Material (Membrane) Ultracel Low-binding Regenerated Cellulose
MWCO 30 kDa
Filtration Area 1 cm2
For Use With (Application) Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment
Housing Material Polypropylene
Includes 12/Pk. Amicon Ultra NMWCO 30 kd, 3mL Protein A resin (Binding capacity: 20mg human IgG/mL agarose), 2 x 66mL Bind/Wash Buffer, 12mL Elution buffer, 2.5mL Neutralization buffer
Max. Force 4000xg for concentration and buffer exchange spin; 1000xg for flow through, wash, elution and reverse spin
Quantity 12/Pk.
Type Amicon Pro Affinity Concentration Kit Protein A with 30 kd Amicon Ultra-0.5 Device
Volume (Metric) 10 mL
Length (English) 4.7 in.
Length (Metric) 12 cm
Diameter (English) 1.2 in.
Diameter (Metric) 29.7 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.