Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ AMPK (α1/β1/γ1), HisTag™, Human, Recombinant Protein, S. frugiperda

Catalog No. 1715365UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-153-65UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-153-65UG Supplier MilliporeSigma™ Supplier No. 1715365UG
Only null left
Add to Cart
Add to Cart
Full length, recombinant, human AMPK subunits (A1, B1, and G1), each fused at the C-terminus to a His·Tag™ sequence and co-expressed in S. frugiperda insect cells using a baculovirus expression system. AMPK is involved in sensing energy levels in the cell that result from changes in AMP levels. AMPK is an upstream regulator of mTOR activation. The active complex is useful in the study of kinase activity regulation and inhibitor screening.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 300mM NaCl, 150mM Imidazole, 50mM Sodium Phosphate, 0.2mM DTT, 0.1mM PMSF, 25% glycerol, pH 7.0.
Purification Method SDS-PAGE
Quantity 5 μg
Source S. frugiperda
Immunogen AMPK (α1/β1/γ1)
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Conjugate His-Tag
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.