Learn More
Description
Specifications
Specifications
| Species | Human |
| Purification Method | HPLC |
| Purity | ≥95% |
| Content And Storage | Reconstitute just prior to use. Do not store following reconstitution, as aggregation may occur. |
| Format | Lyophilized |
| Formulation | Supplied as a chloride salt. |
| For Use With (Application) | Substrate Cleavage Assays (30 to 100μg/mL) |
| Product Type | β-Amyloid Peptide (1-42) |
| Quantity | 0.25 mg |
| Regulatory Status | RUO |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.