Learn More
MilliporeSigma™ Apolipoprotein B, Human Plasma, Low-Density Lipoprotein, Protein
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | Lyophilized from 50mM Na2CO3, 50mM NaCl, 10mM sodium deoxycholate, pH 10.0. |
| Preparation Method | H2O or Aqueous Buffers. Please refer to the label for lot-specific reconstitution volume. |
| Purification Method | SDS-PAGE |
| Quantity | 500 μg |
| Immunogen | Apolipoprotein B |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Solid |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.