Learn More
MilliporeSigma™ Apolipoprotein C-II, Human Plasma, Very Low-Density Lipoprotein, Protein
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | Lyophilized from 10mM NH4HCO3, pH 7.4. |
| Preparation Method | H2O. Please refer to vial label for lot-specific volume prior to lyophilization. |
| Purification Method | SDS-PAGE |
| Quantity | 50 μg |
| Immunogen | Apolipoprotein C-II |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Lyophilized |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.