Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Aquaporin 1 (243-261), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1786111ST Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-861-11ST 1 Set Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-861-11ST Supplier MilliporeSigma™ Supplier No. 1786111ST
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifically detects Aquaporin 1 (243-261) Clone: in Human, Rat samples, and it is validated for Immunohistochemistry (Frozen), Immuno Blotting

AQP1 is a 269 amino acid transmembrane protein belonging to the aquaporin family. AQP1 forms the major water transports pathway of red cells and renal proximal tubules and thin descending limb of Henle, but not collecting duct.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin 1 (243-261)
Applications Immunoblot, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Formulation 50μl antibody lyophilized from PBS, pH 7.4 and 40μg blocking peptide lyophilized from PBS.
Host Species Rabbit
Immunogen a synthetic peptide (KVWTSGQVEEYDLDADDIN) corresponding to amino acids 243-261 of human AQP1, conjugated to KLH
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes aquaporin-1 in red blood cells. Supplied with AQP1243-261 blocking peptide (immunogen).
Includes 50μL antibody lyophilized from PBS, pH 7.4 and 40μg blocking peptide lyophilized from PBS.
Target Species Human, Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.