Learn More
Description
Specifications
Specifications
| Preparation Method | Dilute buffer. enzyme will denature below pH 4.0. |
| Quantity | 1000 U |
| Source | Cucurbita sp. |
| Immunogen | Ascorbate Oxidase |
| Storage Requirements | Following reconstitution refrigerate (4°C). This Stock solutions are stable for up to 3 weeks at 4°C. |
| Regulatory Status | RUO |
| Recombinant | Native |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.