Learn More
MilliporeSigma™ Automatic Sanitization Module for PE tank
Catalog No. TANKASMIN Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
Elix Advantage and Milli-Q Integral Water Purification System
Catalog No. For Use With (Equipment)
TANKASMIN Elix Advantage and Milli-Q Integral Water Purification System
1 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. TANKASMIN Supplier MilliporeSigma™ Supplier No. TANKASMIN
Add to Cart
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
For use with Elix™ Advantage and Milli-Q™ Integral water purification systems
  • For regular emission of UV lightinside the tank to prevent bacterial development
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Elix Advantage and Milli-Q Integral Water Purification System
Type Automatic Sanitization Module
For Use With (Application) Regular emission of UV light inside the Polyethylene tank to prevent bacteria development
Electrical Requirements 230 V, 50 Hz, Single Phase