Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Azure II Staining Dye

Catalog No. M1092110010 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1092110010 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1092110010 Supplier MilliporeSigma™ Supplier No. 1092110010
Add to Cart
Add to Cart

Azure II (C.I.52010/52015) for microscopy. CAS 37247-10-2, pH 2.9 (10g/l, H2O, 20°C).

Azure II (C.I. 52010/ 52015) is a dry dye for the preparation of a color staining solution used for the staining of blood and bone marrow smears of human origin.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 g
For Use With (Application) Staining of blood and bone marrow smears
Product Type Azure Staining Dye
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.