Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ c-MET, GST-Fusion, Human, Recombinant Protein, S. frugiperda

Catalog No. 32588510UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-588-510UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-588-510UG Supplier MilliporeSigma™ Supplier No. 32588510UG
Add to Cart
Add to Cart
Recombinant, human c-MET (amino acids 956-1390) (Genebank entry NM_000245) fused at the N-terminus to a GST-His6-thrombin cleavage site sequence and expressed in S. frugiperda insect cells using a baculovirus expression system. c-Met is a receptor tyrosine kinase activated by hepatocyte growth factor (HGF)/scatter factor (SF). c-MET is a plasminogen-like protein thought to be a humoral mediator of liver regeneration.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 100mM NaCl, 50mM Tris-HCl, 5mM DTT, 4mM reduced glutathione, 20% glycerol, pH 8.0.
Quantity 10 μg
Source S. frugiperda
Immunogen c-MET
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Conjugate GST-Fusion
Species Human
Recombinant Recombinant
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.