Learn More
Description
Specifications
Specifications
| Concentration | Concentration on Label |
| Formulation | In 150mM NaCl, 20mM Tris, 2mM CaCl2, 0.1% NaN3, pH 7.5. |
| Preparation Method | H2O |
| Purification Method | Single band by SDS-PAGE |
| Quantity | 1 mg |
| Immunogen | C-Reactive Protein |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.