Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ CaV2.2 (851-867), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 6815051ST Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
68-150-51ST 1 Set Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-150-51ST Supplier MilliporeSigma™ Supplier No. 6815051ST
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifically detects CaV2.2 (851-867) Clone: in Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunohistochemistry, Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen CaV2.2 (851-867)
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation 50μl antibody lyophilized from PBS, 5% sucrose, 1% BSA, pH. 7.4 and 40μg control peptide lyophilized from PBS.
Host Species Rabbit
Immunogen a synthetic peptide [(C)RHHRHRDRDKTSASTPA] corresponding to amino acids 851-867 of the α1B-subunit of rat brain voltage-gated calcium channels, conjugated to KLH
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽210kDa and the ∽240kDa forms of α1B-subunit of Cav2.2. Supplied with a control peptide.
Includes 50μL antibody lyophilized from PBS, 5% sucrose, 1% BSA, pH. 7.4 and 40μg control peptide lyophilized from PBS.
Target Species Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.