Learn More
Description
- Specific for biotin, biotinylated proteins or antibodies
- Form: Liquid
- Formulation: In 150mM NaCl, 20mM phosphate buffer, 1% BSA, pH 7.6
- Preservative: ≤0.1% sodium azide
Specifications
Specifications
| Quantity | 1 mL |
| Content And Storage | -20°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.