Learn More
MilliporeSigma™ Calbiochem™ Cell Culture Tested Kanamycin Sulfate
Catalog No. 4204115GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-041-15GM 5 g
42-041-125GM 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42-041-15GM Supplier MilliporeSigma™ Supplier No. 4204115GM
An aminoglycoside antibiotic effective against Gram-positive and Gram-negative organisms. Inhibitor of protein biosynthesis that acts on the 30S ribosome, causing misreading of the genetic code.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 15-30°C
Antibiotic Spectrum Gram-Positive and Gram-Negative Bacteria
Quantity 5 g