Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Homodimer Human Recombinant PDGF, BB

Catalog No. 52122510UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-122-510UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-122-510UG Supplier MilliporeSigma™ Supplier No. 52122510UG

Recombinant, human platelet-derived growth factor (disulfide-linked dimer of two 109 amino acid B-chain monomers) expressed in E.Coli

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage ≤-70°C
Formulation Lyophilized from a sterile-filtered solution, carrier free
Quantity 10 μg
Source E. coli
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.