Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Human Recombinant TGF-alpha

Catalog No. 616430100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-643-0100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-643-0100UG Supplier MilliporeSigma™ Supplier No. 616430100UG

Recombinant, human transforming growth factor-α expressed in E. Coli

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage ≤-70°C
Formulation Lyophilized from 30% acetonitrile, 0.1% TFA
Quantity 100 μg
Source E. coli
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.