Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Molecular Biology Grade Glycerol

Catalog No. 3563521L Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-635-21L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 35-635-21L Supplier MilliporeSigma™ Supplier No. 3563521L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polyol (sugar alcohol) compound with three hydroxyl groups

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°-30°C
Product Type Glycerol
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.