Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Mouse Recombinant VEGF164

Catalog No. 6764745UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 μ g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-647-45UG 5 μ g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-647-45UG Supplier MilliporeSigma™ Supplier No. 6764745UG

missing translation for 'validMainframeMsg'

Recombinant, mouse VEGF expressed in S. frugiperda insect cells

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage ≤-70°C
Quantity 5 μ g
Source S. frugiperda insect cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.